3-Bromo-2,4,6-trimethylnitrobenzene ≥95%

Supplier: Matrix Scientific

007063-1G 007063-5G
101791-934EA 70.77 USD
101791-934 101791-936
3-Bromo-2,4,6-trimethylnitrobenzene ≥95%
3-Bromo-2,4,6-trimethylnitrobenzene

Hazard: Irritant/Light Sensitive

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
Boiling Pt: 106…107 °C (1.3 mmHg)
Melting Pt: 64…66 °C
MDL Number: MFCD00015924
CAS Number: 90561-85-6

Order Now

Specification Test Results

Boiling point 106-107°C/1
Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR