1-(4-Trifluoromethylphenyl)-1,2-propandione

Supplier: Matrix Scientific

006902-1G 006902-5G
101791-504EA 64.62 USD
101791-504 101791-506
1-(4-Trifluoromethylphenyl)-1,2-propandione
1-(4-Trifluoromethylphenyl)-1,2-propandione

Hazard: Irritant

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Boiling Pt: 70 °C (0.2 mmHg)
Melting Pt: 30…32 °C
MDL Number: MFCD01941310
CAS Number: 10557-13-8

Order Now

Specification Test Results

Boiling point 70°C/0.2mm
Melting point 30-32°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR