1-(3-Trifluoromethylphenyl)-1,2-propandione

Supplier: Matrix Scientific

Synonyms: 2-Oxo-3'-(trifluoromethyl)propiophenone

006818-1G 006818-5G
101791-210EA 58.46 USD
101791-210 101791-212
1-(3-Trifluoromethylphenyl)-1,2-propandione
1-(3-Trifluoromethylphenyl)-1,2-propandione

Hazard: Irritant

Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Boiling Pt: 80 °C (0.1 mmHg)
Melting Pt: 25…26 °C
MDL Number: MFCD01941304
CAS Number: 10557-15-0

Order Now

Specification Test Results

Boiling point 80°C/0.1mm
Melting point 25-26°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR