1-(4-Chlorophenyl)-1,2-propandione

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)propane-1,2-dione

006816-1G 006816-5G
101791-202EA 347.69 USD
101791-202 101791-204
1-(4-Chlorophenyl)-1,2-propandione
1-(4-Chlorophenyl)-1,2-propandione

Hazard: Irritant

Formula: C₉H₇ClO₂
MW: 182.61 g/mol
Boiling Pt: 60-65°C/0.02mm
Melting Pt: 30…32 °C
MDL Number: MFCD01941300
CAS Number: 10557-21-8

Order Now

Specification Test Results

Boiling point 60-65°C/0.0
Melting point 30-32°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR