2-(Trifluoromethyl)pyridine-5-carboxaldehyde

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)nicotinaldehyde

006813-1G 006813-5G
101791-192EA 41.54 USD
101791-192 101791-194
2-(Trifluoromethyl)pyridine-5-carboxaldehyde
2-(Trifluoromethyl)pyridine-5-carboxaldehyde

Hazard: Irritant

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Boiling Pt: 72…74 °C (0.6 mmHg)
Melting Pt: 53…55 °C
MDL Number: MFCD01862647
CAS Number: 386704-12-7

Order Now

Specification Test Results

Boiling point 72-74°C/0.6
Melting point 53-55°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR