4-(Dimethylamino)phenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

006622-5G 006622-25G
101790-826EA 44.62 USD
101790-826 101790-828
4-(Dimethylamino)phenyl isothiocyanate ≥98%
4-(Dimethylamino)phenyl isothiocyanate

Hazard: Toxic/Corrosive/Stored under Argon/Refrigerate/Lachrymator

Formula: C₉H₁₀N₂S
MW: 178.25 g/mol
Boiling Pt: 151…152 °C (6 mmHg)
Melting Pt: 69…71 °C
MDL Number: MFCD00041093
CAS Number: 2131-64-8

Order Now

Specification Test Results

Boiling point 151-152°C/6
Melting point 69-71°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR