3-[3,5-Bis(trifluoromethyl)phenyl]prop-2-yn-1-ol
Supplier: Matrix Scientific
006182-250MG
006182-1G
101790-212EA
89.23
USD
101790-212
101790-214
3-[3,5-Bis(trifluoromethyl)phenyl]prop-2-yn-1-ol
3-[3,5-Bis(trifluoromethyl)phenyl]prop-2-yn-1-ol
Hazard: Irritant
Formula:
C₁₁H₆F₆O MW: 268.16 g/mol Boiling Pt: 84 °C (1 mmHg) Melting Pt: 58…60 °C |
MDL Number:
MFCD00052418 CAS Number: 81613-61-8 |
Specification Test Results
Boiling point | 84°C/1mm |
Melting point | 58-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...