3-(4-Chlorophenyl)propiononitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)propanenitrile

006114-1G 006114-5G
101790-126EA 106.15 USD
101790-126 101790-128
3-(4-Chlorophenyl)propiononitrile ≥98%
3-(4-Chlorophenyl)propiononitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₈ClN
MW: 165.6195 g/mol
Boiling Pt: 120 °C (1 mmHg)
Melting Pt: 25…27 °C
MDL Number: MFCD01310802
CAS Number: 32327-71-2

Order Now

Specification Test Results

Boiling point 120°C/1mm
Melting point 25-27°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR