(3,4-Diethoxyphenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

005670-1G 005670-5G
101789-324EA 138.46 USD
101789-324 101789-326
(3,4-Diethoxyphenyl)acetonitrile ≥98%
(3,4-Diethoxyphenyl)acetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₂H₁₅NO₂
Boiling Pt: 130 °C (1 mmHg)
Melting Pt: 33…35 °C
MDL Number: MFCD00060715
CAS Number: 27472-21-5

Order Now

Specification Test Results

Boiling point 130°C/1mm
Melting point 33-35°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR