4-(Trifluoromethylthio)phenol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-[(Trifluoromethyl)sulphanyl]phenol

005541-5G 005541-25G
101789-098EA 13.85 USD
101789-098 101789-100
4-(Trifluoromethylthio)phenol ≥97%
4-(Trifluoromethylthio)phenol

Hazard: Irritant/Stench

Formula: C₇H₅F₃OS
Boiling Pt: 77-78°C/7mm
Melting Pt: 56-57°C
MDL Number: MFCD00036035
CAS Number: 461-84-7

Order Now

Specification Test Results

Boiling point 77-78°C/7mm
Melting point 56-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR