4-(Trifluoromethyl)benzhydrol ≥97%

Supplier: Matrix Scientific

Synonyms: Phenyl(4-(trifluoromethyl)phenyl)methanol

005460-1G 005460-5G
101788-988EA 29.23 USD
101788-988 101788-990
4-(Trifluoromethyl)benzhydrol ≥97%
4-(Trifluoromethyl)benzhydrol

Hazard: Irritant

Formula: C₁₄H₁₁F₃O
Boiling Pt: 122…124 °C (0.4 mmHg)
Melting Pt: 59…61 °C
MDL Number: MFCD00014399
CAS Number: 395-23-3

Order Now

Specification Test Results

Boiling point 122-124°C/0
Melting point 59-61°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR