5-(Trifluoromethoxy)salicylaldehyde ≥95%
Supplier: Matrix Scientific
005369-1G
005369-5G
005369-25G
101788-766EA
32.31
USD
101788-766
101788-768
102550-862
5-(Trifluoromethoxy)salicylaldehyde ≥95%
5-(Trifluoromethoxy)salicylaldehyde
Hazard: Irritant
Formula:
C₈H₅F₃O₃ Boiling Pt: 82°C/60mm Melting Pt: 31-33°C |
MDL Number:
MFCD00075249 CAS Number: 93249-62-8 |
Specification Test Results
Boiling point | 82°C/60mm |
Melting point | 31-33°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...