tert-Butyl-N-allylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

023768-5G 023768-25G
101816-682EA 49.23 USD
101816-682 101816-684
tert-Butyl-N-allylcarbamate ≥97%
tert-Butyl-N-allylcarbamate

Hazard: Irritant/Refrigerate

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 48…50 °C (0.3 mmHg)
Melting Pt: 36…38 °C
MDL Number: MFCD00191870
CAS Number: 78888-18-3

Order Now

Specification Test Results

Boiling point 48-50°C/0.3
Melting point 36-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR