1,3-Dichloro-2-iodobenzene ≥98%

Supplier: Matrix Scientific

004998-25G 004998-100G
101788-130EA 89.23 USD
101788-130 101788-132
1,3-Dichloro-2-iodobenzene ≥98%
1,3-Dichloro-2-iodobenzene

Hazard: Irritant/Light Sensitive

Formula: C₆H₃Cl₂I
MW: 272.90 g/mol
Boiling Pt: 79°C/1mm
Melting Pt: 62-64°C
MDL Number: MFCD00060658
CAS Number: 19230-28-5

Order Now

Specification Test Results

Boiling point 79°C/1mm
Melting point 62-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR