1,4-Diiodo-2-fluorobenzene ≥99%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-1,4-diiodobenzene
004876-5G
004876-25G
101787-944EA
30.77
USD
101787-944
101787-946
1,4-Diiodo-2-fluorobenzene ≥99%
1,4-Diiodo-2-fluorobenzene
Hazard: Irritant
Formula:
C₆H₃FI₂ MW: 347.9000 g/mol Boiling Pt: 135 °C (20 mmHg) Melting Pt: 69…71 °C |
MDL Number:
MFCD00061107 CAS Number: 147808-02-4 |
Specification Test Results
Boiling point | 135°C/20mm |
Melting point | 69-71°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...