Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol ≥97%
Supplier: Matrix Scientific
004775-5G
004775-25G
101787-794EA
141.54
USD
101787-794
101787-796
Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol ≥97%
Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol
Hazard: Irritant
Formula:
C₆H₂F₁₂O₂ Boiling Pt: 113…114 °C Melting Pt: 18…20 °C |
MDL Number:
MFCD00427700 CAS Number: 918-21-8 |
Specification Test Results
Boiling point | 113-114°C |
Melting point | 18-20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...