4-Nitrobenzoyl isothiocyanate

Supplier: Matrix Scientific

004753-5G 004753-25G
101787-774EA 92.31 USD
101787-774 101787-776
4-Nitrobenzoyl isothiocyanate
4-Nitrobenzoyl isothiocyanate

Hazard: Refrigerate/Lachrymator

Formula: C₈H₄N₂O₃S
Boiling Pt: 187 °C (10 mmHg)
Melting Pt: 90…92 °C
MDL Number: MFCD00041252
CAS Number: 28115-92-6

Order Now

Specification Test Results

Boiling point 187°C/10mm
Melting point 90-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR