1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine ≥97%
Supplier: Matrix Scientific
003756-5G
003756-25G
101786-176EA
21.54
USD
101786-176
101786-178
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine ≥97%
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
Hazard: Irritant
Formula:
C₁₀H₁₂F₃N₃ Boiling Pt: 122°C/2mm Melting Pt: 43…46 °C |
MDL Number:
MFCD00114708 CAS Number: 132834-58-3 |
Specification Test Results
Boiling point | 122°C/2mm |
Melting point | 43-46°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...