2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Hydroxy-2,3,5,6-tetrafluorobenzotrifluoride
003508-5G
003508-25G
101785-810EA
60
USD
101785-810
101785-812
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol
Hazard: Irritant/Irritant-Harmful
Formula:
C₇HF₇O Boiling Pt: 91°C/70mm Melting Pt: 25°C |
MDL Number:
MFCD00155984 CAS Number: 2787-79-3 |
Specification Test Results
Boiling point | 91°C/70mm |
Melting point | 25°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...