2,3,5,6-Tetrafluoro-p-xylene ≥98%
Supplier: Matrix Scientific
003485-5G
003485-25G
101785-770EA
487.69
USD
101785-770
101785-772
2,3,5,6-Tetrafluoro-p-xylene ≥98%
2,3,5,6-Tetrafluoro-p-xylene
Hazard: Irritant
Formula:
C₈H₆F₄ Boiling Pt: 143…144 °C Melting Pt: 35…37 °C |
MDL Number:
MFCD00012231 CAS Number: 703-87-7 |
Specification Test Results
Boiling point | 143-144°C |
Melting point | 35-37°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...