Ethyl-2-naphthoate ≥99%

Supplier: Matrix Scientific

002985-5G 002985-25G
101785-004EA 56.92 USD
101785-004 101785-006
Ethyl-2-naphthoate ≥99%
Ethyl-2-naphthoate

Hazard: Irritant

Formula: C₁₃H₁₂O₂
MW: 200.23 g/mol
Boiling Pt: 308-309°C
Melting Pt: 32°C
MDL Number: MFCD00016811
CAS Number: 3007-91-8

Order Now

Specification Test Results

Boiling point 308-309°C
Melting point 32°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR