1H,1H,9H-Perfluorononyl-p-toluenesulfonate ≥95%

Supplier: Matrix Scientific

002867-25G 002867-100G
101784-806EA 160 USD
101784-806 101784-808
1H,1H,9H-Perfluorononyl-p-toluenesulfonate ≥95%
1H,1H,9H-Perfluorononyl-p-toluenesulfonate

Hazard: Irritant

Formula: C₁₆H₁₀F₁₆O₃S
Boiling Pt: 171.5 °C (2.3 mmHg)
Melting Pt: 46.5 °C
MDL Number: MFCD00085895
CAS Number: 864-23-3

Order Now

Specification Test Results

Boiling point 171.5°C/2.3
Melting point 46.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR