2,2-Bis(4-methylphenyl)hexafluoropropane ≥98%

Supplier: Matrix Scientific

008811-5G 008811-25G
101795-504EA 53.85 USD
101795-504 101795-506
2,2-Bis(4-methylphenyl)hexafluoropropane ≥98%
2,2-Bis(4-methylphenyl)hexafluoropropane

Hazard: Irritant

Formula: C₁₇H₁₄F₆
Boiling Pt: 117 °C (2 mmHg)
Melting Pt: 82…83 °C
MDL Number: MFCD00042597
CAS Number: 1095-77-8

Order Now

Specification Test Results

Boiling point 117°C/2mm
Melting point 82-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR