2,2-Bis(3,4-dimethylphenyl)hexafluoropropane ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4'-(Hexafluoroisopropylidene)di-o-xylene, 4,4'-(Perfluoropropane-2,2-diyl)bis(1,2-dimethylbenzene)

008804-5G 008804-25G
101795-486EA 44.62 USD
101795-486 101795-488
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane ≥97%
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane

Hazard: Irritant

Formula: C₁₉H₁₈F₆
MW: 360.34 g/mol
Boiling Pt: 110…120 °C (2 mmHg)
Melting Pt: 76…78 °C
MDL Number: MFCD00042167
CAS Number: 65294-20-4

Order Now

Specification Test Results

Boiling point 110-120°C/2
Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR