1-(5-Methyl-4-isoxazolyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

044828-500MG 044828-1G 044828-5G
101867-784EA 315.38 USD
101867-784 101867-786 101867-788
1-(5-Methyl-4-isoxazolyl)-1-ethanone ≥95%
1-(5-Methyl-4-isoxazolyl)-1-ethanone

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 54 °C (0.25 mmHg)
Melting Pt: 26…28 °C
MDL Number: MFCD01935998
CAS Number: 6497-21-8

Order Now

Specification Test Results

Boiling point 54°C/0.25mm
Melting point 26-28°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR