α-Methyl-2-trifluoromethylbenzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[2-(Trifluoromethyl)phenyl]ethanol

002213-5G 002213-25G
101783-472EA 52.31 USD
101783-472 101783-474
α-Methyl-2-trifluoromethylbenzyl alcohol ≥97%
alpha-Methyl-2-trifluoromethylbenzyl alcohol

Hazard: Irritant

Formula: C₉H₉F₃O
Boiling Pt: 114…115 °C (2 mmHg)
Melting Pt: 40…42 °C
MDL Number: MFCD00004513
CAS Number: 79756-81-3

Order Now

Specification Test Results

Boiling point 114-115°C/2
Melting point 40-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR