4-Chloro-2-methylbenzonitrile ≥98%

Supplier: Matrix Scientific

002177-5G 002177-25G
101783-394EA 61.54 USD
101783-394 101783-396
4-Chloro-2-methylbenzonitrile ≥98%
4-Chloro-2-methylbenzonitrile

Hazard: Irritant-Harmful

Formula: C₈H₆ClN
MW: 151.60 g/mol
Boiling Pt: 85…86 °C (3 mmHg)
Melting Pt: 56…58 °C
MDL Number: MFCD00045603
CAS Number: 50712-68-0

Order Now

Specification Test Results

Boiling point 85-86°C/3mm
Melting point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR