3-Chloro-2-methylbenzonitrile ≥98%

Supplier: Matrix Scientific

002175-1G 002175-5G
101783-390EA 23.08 USD
101783-390 101783-392
3-Chloro-2-methylbenzonitrile ≥98%
3-Chloro-2-methylbenzonitrile

Hazard: Irritant-Harmful

Formula: C₈H₆ClN
MW: 151.60 g/mol
Boiling Pt: 120°C/22mm
Melting Pt: 21-23°C
MDL Number: MFCD00045598
CAS Number: 54454-12-5

Order Now

Specification Test Results

Boiling point 120°C/22mm
Melting point 21-23°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR