5-Chloro-2-methylbenzonitrile ≥98%
Supplier: Matrix Scientific
002170-5G
002170-25G
101783-374EA
143.08
USD
101783-374
101783-376
5-Chloro-2-methylbenzonitrile ≥98%
5-Chloro-2-methylbenzonitrile
Hazard: Irritant-Harmful
Formula:
C₈H₆ClN MW: 151.5929 g/mol Boiling Pt: 86 °C (3 mmHg) Melting Pt: 43…45 °C |
MDL Number:
MFCD00045599 CAS Number: 50712-70-4 |
Specification Test Results
Boiling point | 86°C/3mm |
Melting point | 43-45°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...