1H,1H,11H-Eicosafluoroundecyl acrylate ≥92% inhibited

Supplier: Matrix Scientific


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 50 ppm

001535-25G 001535-100G
101781-916EA 280 USD
101781-916 101781-918
1H,1H,11H-Eicosafluoroundecyl acrylate ≥92% inhibited
1H,1H,11H-Eicosafluoroundecyl acrylate

Hazard: Irritant/Refrigerate

Formula: C₁₄H₆F₂₀O₂
Boiling Pt: 115 °C (1 mmHg)
Melting Pt: 44…45 °C
MDL Number: MFCD00042112
CAS Number: 4998-38-3

Order Now

Specification Test Results

Boiling point 115°C/1mm
Melting point 44-45°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR