4-Hydroxy-3-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(4-Hydroxy-3-methoxyphenyl)acetonitrile

001458-1G 001458-5G
101781-708EA 130.77 USD
101781-708 101781-710
4-Hydroxy-3-methoxyphenylacetonitrile ≥98%
4-Hydroxy-3-methoxyphenylacetonitrile

Hazard: Irritant-Harmful

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 135…145 °C (2 mmHg)
Melting Pt: 56…57 °C
MDL Number: MFCD00001920
CAS Number: 4468-59-1

Order Now

Specification Test Results

Boiling point 135-145°C/2
Melting point 56-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR