1H,1H,7H-Dodecafluoroheptyl-p-toluenesulfonate ≥95%

Supplier: Matrix Scientific

002821-5G 002821-25G
101784-718EA 76.92 USD
101784-718 101784-720
1H,1H,7H-Dodecafluoroheptyl-p-toluenesulfonate ≥95%
1H,1H,7H-Dodecafluoroheptyl-p-toluenesulfonate

Hazard: Irritant

Formula: C₁₄H₁₀F₁₂O₃S
MW: 486.28 g/mol
Boiling Pt: 126 °C (0.5 mmHg)
Melting Pt: 32 °C
MDL Number: MFCD00490197
CAS Number: 424-16-8

Order Now

Specification Test Results

Boiling point 126°C/0.5mm
Melting point 32°C
ND20 1.4123

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR