4-Methoxythioanisole ≥97%
Supplier: Matrix Scientific
001483-25G
001483-100G
101781-794EA
224.62
USD
101781-794
101781-796
4-Methoxythioanisole ≥97%
4-Methoxythioanisole
Hazard: Irritant
Formula:
C₈H₁₀OS MW: 154.2294 g/mol Boiling Pt: 99°C/4mm Melting Pt: 22-23°C |
MDL Number:
MFCD00010587 CAS Number: 1879-16-9 |
Specification Test Results
Boiling point | 99°C/4mm |
Melting point | 22-23°C |
ND20 | 1.5795 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...