Perfluoro(2-methyl-3-pentanone) ≥98%

Supplier: Matrix Scientific

Synonyms: 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Perfluoroethyl Perfluoroisopropyl ketone

009117-5G 009117-25G
101796-400EA 49.23 USD
101796-400 101796-402
Perfluoro(2-methyl-3-pentanone) ≥98%
Perfluoro(2-methyl-3-pentanone)

TCA listed: T
Hazard: Irritant

Formula: C₆F₁₂O
MW: 316.05 g/mol
Boiling Pt: 49°C
Melting Pt: –108 °C
Density: 1.6
MDL Number: MFCD04038343
CAS Number: 756-13-8

Order Now

Specification Test Results

Boiling point 49°C
Melting point -108°C
Density 1.6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR