1-Chloro-2,2,3,3-tetrafluoroethane ≥97%
Supplier: Matrix Scientific
008880-1G
008880-5G
101795-704EA
63.08
USD
101795-704
101795-706
1-Chloro-2,2,3,3-tetrafluoroethane ≥97%
1-Chloro-2,2,3,3-tetrafluoroethane
Hazard: Ozone Depleter/Irritant
Formula:
C₂HClF₄ Boiling Pt: 53-54°C Melting Pt: -101.8°C Density: 1.446 |
CAS Number:
679-85-6 |
Specification Test Results
Boiling point | 53-54°C |
Melting point | -101.8°C |
Density | 1.446 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...