2-Bromo-4,5-difluorobenzonitrile ≥98%

Supplier: Matrix Scientific

067254-1G 067254-5G 067254-25G
101933-638EA 183.08 USD
101933-638 101933-640 101933-642
2-Bromo-4,5-difluorobenzonitrile ≥98%
2-Bromo-4,5-difluorobenzonitrile

Hazard: Irritant

Formula: C₇H₂BrF₂N
Boiling Pt: 255.5°C
Melting Pt: 63-64°C
Density: 1.774
Flash Pt: 108°C
MDL Number: MFCD09864661
CAS Number: 64695-82-5

Order Now

Specification Test Results

Boiling point 255.5°C
Melting point 63-64°C
Density 1.774


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR