2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane ≥97%

Supplier: Matrix Scientific

005085-1G 005085-5G
101788-244EA 96.92 USD
101788-244 101788-246
2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane ≥97%
2H-Perfluoro-5,8-dimethyl-3,6,9-trioxadodecane

TCA listed: T
Hazard: Irritant

Formula: C₁₁HF₂₃O₃
Boiling Pt: 151-154°C
Melting Pt: –106.7 °C
Density: 1.72
MDL Number: MFCD00054715
CAS Number: 3330-16-3

Order Now

Specification Test Results

Boiling point 151-154°C
Melting point -106.7°C
Density 1.72
ND20 1.2678


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR