2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3-Tetrafluoropropyl 4-methylbenzenesulphonate

003479-5G 003479-25G
101785-762EA 1373.85 USD
101785-762 101785-764
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate ≥97%
2,2,3,3-Tetrafluoropropyl-p-toluenesulfonate

Hazard: Irritant

Formula: C₁₀H₁₀F₄O₃S
MW: 286.25 g/mol
Boiling Pt: 124 °C (2 mmHg)
Melting Pt: 13…16 °C
Density: 1.397
MDL Number: MFCD00155960
CAS Number: 786-31-2

Order Now

Specification Test Results

Boiling point 124°C/2mm
Melting point 13-16°C
Density 1.397
ND20 1.4602

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR