2H-Perfluoro-5-methyl-3,6-dioxanonane ≥97%

Supplier: Matrix Scientific

Synonyms: 1,1,1,2,2,3,3-Heptafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy]propane

009115-1G 009115-5G
101796-392EA 43.08 USD
101796-392 101796-394
2H-Perfluoro-5-methyl-3,6-dioxanonane ≥97%
2H-Perfluoro-5-methyl-3,6-dioxanonane

Hazard: Irritant

Formula: C₈HF₁₇O₂
MW: 452.07 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –122 °C
Density: 1.659
MDL Number: MFCD00054714
CAS Number: 3330-14-1

Order Now

Specification Test Results

Boiling point 103-105°C
Melting point -122°C
Density 1.659
ND20 1.2598

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR