1,2-Dichlorotetrafluoropropene ≥97%

Supplier: Matrix Scientific

008908-1G 008908-5G
101795-788EA 89.23 USD
101795-788 101795-790
1,2-Dichlorotetrafluoropropene ≥97%
1,2-Dichlorotetrafluoropropene

Hazard: Irritant

Formula: C₃Cl₂F₄
Boiling Pt: 47.3 °C
Melting Pt: –137 °C
Density: 1.5468
MDL Number: MFCD04038281
CAS Number: 431-53-8

Order Now

Specification Test Results

Boiling point 47.3°C
Melting point -137°C
Density 1.5468
ND20 1.3511


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR