Methoxyflurane ≥98%

Supplier: Matrix Scientific

Synonyms: 2,2-Dichloro-1,1-difluoroethyl methyl ether

008896-1G 008896-5G
101795-748EA 49.23 USD
101795-748 101795-750
Methoxyflurane ≥98%
Methoxyflurane

Hazard: Flammable

Formula: C₃H₄Cl₂F₂O
MW: 164.961 g/mol
Boiling Pt: 103…104.5 °C
Melting Pt: –35 °C
Density: 1.4262
MDL Number: MFCD00040144
CAS Number: 76-38-0

Order Now

Specification Test Results

Boiling point 103-104.5°C
Melting point -35°C
Density 1.4262
ND20 1.3861

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR