3-(Trifluoromethyl)quinoxalin-2-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-3-(trifluoromethyl)quinoxaline, 3-(Trifluoromethyl)quinoxalin-2(1H)-one

070808-500MG 070808-1G 070808-5G
101946-408EA 193.85 USD
101946-408 101946-410 101946-412
3-(Trifluoromethyl)quinoxalin-2-ol ≥95%
3-(Trifluoromethyl)quinoxalin-2-ol

Hazard: Irritant

Formula: C₉H₅F₃N₂O
MW: 214.15 g/mol
Melting Pt: 248…249 °C
MDL Number: MFCD00243991
CAS Number: 58457-64-0

Order Now

Specification Test Results

Melting point 248-249°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR