(2E)-3-{1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoic acid ≥95%

Supplier: Matrix Scientific

070801-500MG 070801-1G
101946-386EA 600 USD
101946-386 101946-388
(2E)-3-{1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoic acid ≥95%
(2E)-3-{1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrol-2-yl}prop-2-enoic acid

Hazard: Irritant

Formula: C₁₆H₁₅NO₃
Melting Pt: 188…190 °C
MDL Number: MFCD21333203
CAS Number: 117378-89-9

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR