1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrole-2-carbaldehyde ≥97%
Supplier: Matrix Scientific
070800-500MG
070800-1G
101946-382EA
298.46
USD
101946-382
101946-384
1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrole-2-carbaldehyde ≥97%
1-Methyl-4-[(4-methylphenyl)carbonyl]-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂ Melting Pt: 118…120 °C |
MDL Number:
MFCD21333201 CAS Number: 117378-81-1 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...