Methyl-3-methoxy-4-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methoxy-4-methylbenzoate

070798-500MG 070798-1G 070798-5G
101946-370EA 193.85 USD
101946-370 101946-372 101946-374
Methyl-3-methoxy-4-methylbenzoate ≥95%
Methyl 3-methoxy-4-methylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Melting Pt: 42…46 °C
MDL Number: MFCD00082710
CAS Number: 3556-83-0

Order Now

Specification Test Results

Melting point 42-46°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR