3-Methyl-2-quinoxalinol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methylquinoxalin-2-ol

070797-500MG 070797-1G 070797-5G
101946-364EA 193.85 USD
101946-364 101946-366 101946-368
3-Methyl-2-quinoxalinol ≥95%
3-Methyl-2-quinoxalinol

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.176 g/mol
Melting Pt: 248…249 °C
MDL Number: MFCD00047567
CAS Number: 14003-34-0

Order Now

Specification Test Results

Melting point 248-249°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR