Methyl-2-bromo-4-fluoro-5-methoxybenzoate ≥95%

Supplier: Matrix Scientific

070796-500MG 070796-1G
101946-360EA 261.54 USD
101946-360 101946-362
Methyl-2-bromo-4-fluoro-5-methoxybenzoate ≥95%
Methyl-2-bromo-4-fluoro-5-methoxybenzoate

Hazard: Irritant

Formula: C₉H₈BrFO₃
Melting Pt: 36…38 °C
MDL Number: MFCD20482372
CAS Number: 1007455-22-2

Order Now

Specification Test Results

Melting point 36-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR