(3-Methoxy-2-methylphenyl)methanol ≥95%

Supplier: Matrix Scientific

070793-500MG 070793-1G
101946-350EA 403.08 USD
101946-350 101946-352
(3-Methoxy-2-methylphenyl)methanol ≥95%
(3-Methoxy-2-methylphenyl)methanol

Hazard: Irritant

Formula: C₉H₁₂O₂
Melting Pt: 63…66 °C
MDL Number: MFCD12031988
CAS Number: 33797-34-1

Order Now

Specification Test Results

Melting point 63-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR