Ethyl-3-(trifluoromethyl)quinoxaline-2-carboxylate ≥95%

Supplier: Matrix Scientific

070788-500MG 070788-1G
101946-330EA 600 USD
101946-330 101946-332
Ethyl-3-(trifluoromethyl)quinoxaline-2-carboxylate ≥95%
Ethyl-3-(trifluoromethyl)quinoxaline-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD08690765
CAS Number: 3885-40-3

Order Now

Specification Test Results

Melting point 47-49°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR