Ethyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

070786-500MG 070786-1G
101946-324EA 529.23 USD
101946-324 101946-326
Ethyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate ≥95%
Ethyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₂
Melting Pt: 213…215 °C (dec.)
MDL Number: MFCD13195432
CAS Number: 915142-91-5

Order Now

Specification Test Results

Melting point 213-215°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR